Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sun5
Entrez Gene ID
100759609
Gene Name
Sad1 and UNC84 domain containing 5
Gene Aliases
I79_008860
Chromosome Location
Chr.Un|NW_003613890: 162511 - 176911 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502338.1 XP_003502386.1 9 - 10 645 71

View Full Product Details