Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pwwp2b
Entrez Gene ID
100772421
Gene Name
PWWP domain containing 2B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617306: 61176 - 67399 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515077.2 XP_003515125.1 2 - 3 260 66

View Full Product Details