Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il4i1
Entrez Gene ID
100773063
Gene Name
interleukin 4 induced 1
Gene Aliases
I79_019299
Chromosome Location
Chr.Un|NW_003614984: 101263 - 109876 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651465.1 XP_007649655.1 5 - 6 755 68
XM_007651466.1 XP_007649656.1 5 - 6 755 68

View Full Product Details