Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc34
Entrez Gene ID
100755808
Gene Name
tetratricopeptide repeat domain 34
Gene Aliases
I79_022520
Chromosome Location
Chr.Un|NW_003615985: 96858 - 126551 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653796.1 XP_007651986.1 4 - 5 1211 100

View Full Product Details