Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcer2
Entrez Gene ID
100769597
Gene Name
Fc fragment of IgE, low affinity II, receptor for (CD23)
Gene Aliases
I79_015483
Chromosome Location
Chr.Un|NW_003614420: 564766 - 574025 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507751.2 XP_003507799.1 7 - 8 449 57
XM_007648348.1 XP_007646538.1 7 - 8 446 57
XM_007648349.1 XP_007646539.1 6 - 7 383 57

View Full Product Details