Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl33
Entrez Gene ID
100756498
Gene Name
mitochondrial ribosomal protein L33
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613628: 2826499 - 2834470 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496574.2 XP_003496622.1 2 - 3 272 69

View Full Product Details