Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dfnb59
Entrez Gene ID
100766395
Gene Name
deafness, autosomal recessive 59
Gene Aliases
I79_007371, pejvakin
Chromosome Location
Chr.Un|NW_003613827: 550367 - 557448 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501022.2 XP_003501070.1 2 - 3 588 90

View Full Product Details