Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nkrf
Entrez Gene ID
100764046
Gene Name
NFKB repressing factor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613768: 450083 - 465663 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500016.2 XP_003500064.1 2 - 3 154 89

View Full Product Details