Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pnma2
Entrez Gene ID
100759435
Gene Name
paraneoplastic Ma antigen 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614124: 1099596 - 1100690 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505163.1 XP_003505211.1 1 - 1 1015 66

View Full Product Details