Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
N4bp1
Entrez Gene ID
100750417
Gene Name
NEDD4 binding protein 1
Gene Aliases
I79_016205
Chromosome Location
Chr.Un|NW_003614520: 598154 - 640209 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648941.1 XP_007647131.1 1 - 2 499 62

View Full Product Details