Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppa1
Entrez Gene ID
100769185
Gene Name
pyrophosphatase (inorganic) 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613759: 1023127 - 1051336 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640632.1 XP_007638822.1 9 - 10 939 59
GenBank mRNA JI865597.1 - 9 - 10 909 59

View Full Product Details