Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hrh4
Entrez Gene ID
100774770
Gene Name
histamine receptor H4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614049: 132244 - 150882 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504279.2 XP_003504327.2 1 - 2 192 99

View Full Product Details