Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Emilin3
Entrez Gene ID
100771706
Gene Name
elastin microfibril interfacer 3
Gene Aliases
EMILIN-3, I79_015404
Chromosome Location
Chr.Un|NW_003614415: 483951 - 493109 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648277.1 XP_007646467.1 3 - 4 508 103

View Full Product Details