Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Otos
Entrez Gene ID
100770042
Gene Name
otospiralin
Gene Aliases
I79_005295
Chromosome Location
Chr.Un|NW_003613726: 248500 - 249538 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499252.1 XP_003499300.1 2 - 3 90 108

View Full Product Details