Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc38a8
Entrez Gene ID
100750441
Gene Name
solute carrier family 38, member 8
Gene Aliases
I79_000278
Chromosome Location
Chr.Un|NW_003613583: 4057517 - 4076225 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495123.1 XP_003495171.1 1 - 2 189 55

View Full Product Details