Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Barhl1
Entrez Gene ID
100774052
Gene Name
BarH-like homeobox 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616009: 96396 - 104049 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653837.1 XP_007652027.1 3 - 4 910 67

View Full Product Details