Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Leprel2
Entrez Gene ID
100767056
Gene Name
leprecan-like 2
Gene Aliases
I79_001409
Chromosome Location
Chr.Un|NW_003613610: 3865854 - 3879883 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646914.1 XP_007645104.1 13 - 14 1443 64
GenBank mRNA JI865657.1 - 1 - 2 56 64

View Full Product Details