Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rad51ap2
Entrez Gene ID
100757413
Gene Name
RAD51 associated protein 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613936: 483788 - 493054 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503049.1 XP_003503097.1 3 - 4 2745 104

View Full Product Details