Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kank3
Entrez Gene ID
103160001
Gene Name
KN motif and ankyrin repeat domains 3
Gene Aliases
I79_018816
Chromosome Location
Chr.Un|NW_003614895: 386389 - 391618 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651135.1 XP_007649325.1 4 - 5 765 62

View Full Product Details