Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc34
Entrez Gene ID
100752060
Gene Name
coiled-coil domain containing 34
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614939: 158599 - 183790 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510697.2 XP_003510745.1 2 - 3 533 119
GenBank mRNA JI878551.1 - 4 - 5 119
JP044007.1 - 2 - 3 528 119

View Full Product Details