Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cacng4
Entrez Gene ID
100771968
Gene Name
calcium channel, voltage-dependent, gamma subunit 4
Gene Aliases
I79_008283
Chromosome Location
Chr.Un|NW_003613864: 244990 - 269846 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501870.2 XP_003501918.1 2 - 3 298 86

View Full Product Details