Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il17f
Entrez Gene ID
100751004
Gene Name
interleukin 17F
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614654: 381936 - 398129 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649727.1 XP_007647917.1 3 - 4 455 107
XM_007649729.1 XP_007647919.1 4 - 5 471 107
XM_007649730.1 XP_007647920.1 1 - 2 236 107

View Full Product Details