Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Icam5
Entrez Gene ID
100772237
Gene Name
intercellular adhesion molecule 5, telencephalin
Gene Aliases
I79_005887
Chromosome Location
Chr.Un|NW_003613752: 1182785 - 1191203 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499772.2 XP_003499820.1 10 - 11 3784 54

View Full Product Details