Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il17rb
Entrez Gene ID
100753686
Gene Name
interleukin 17 receptor B
Gene Aliases
I79_001465
Chromosome Location
Chr.Un|NW_003613611: 3572478 - 3584424 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496128.1 XP_003496176.1 7 - 8 670 60

View Full Product Details