Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcdc1
Entrez Gene ID
100769645
Gene Name
doublecortin domain containing 1
Gene Aliases
I79_003164
Chromosome Location
Chr.Un|NW_003613653: 1767527 - 1977672 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007614116.1 XP_007612306.1 9 - 10 1364 80

View Full Product Details