Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss21
Entrez Gene ID
100770108
Gene Name
protease, serine, 21 (testisin)
Gene Aliases
I79_022426, Testisin
Chromosome Location
Chr.Un|NW_003615934: 9364 - 14463 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513449.2 XP_003513497.1 1 - 2 127 63

View Full Product Details