Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spocd1
Entrez Gene ID
100773782
Gene Name
SPOC domain containing 1
Gene Aliases
I79_004145
Chromosome Location
Chr.Un|NW_003613689: 17909 - 48499 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007627072.1 XP_007625262.1 5 - 6 1113 83

View Full Product Details