Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nus1
Entrez Gene ID
100761477
Gene Name
nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae)
Gene Aliases
I79_014547
Chromosome Location
Chr.Un|NW_003614311: 561731 - 592305 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647508.1 XP_007645698.1 6 - 7 881 88
GenBank mRNA JP045610.1 - 4 - 5 494 88

View Full Product Details