Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtn2
Entrez Gene ID
100756065
Gene Name
reticulon 2
Gene Aliases
I79_012659
Chromosome Location
Chr.Un|NW_003614150: 651938 - 664973 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645973.1 XP_007644163.1 5 - 6 949 68
GenBank mRNA JI879304.1 - 1 - 2 144 68

View Full Product Details