Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekhf2
Entrez Gene ID
100760693
Gene Name
pleckstrin homology domain containing, family F (with FYVE domain) member 2
Gene Aliases
I79_010286
Chromosome Location
Chr.Un|NW_003613974: 822201 - 827216 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503480.2 XP_003503528.1 1 - 2 77 77

View Full Product Details