Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pold2
Entrez Gene ID
100762815
Gene Name
polymerase (DNA directed), delta 2, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614085: 355887 - 361981 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504751.2 XP_003504799.2 9 - 10 1244 61
XM_007645263.1 XP_007643453.1 9 - 10 1694 61
XM_007645264.1 XP_007643454.1 9 - 10 1886 61
GenBank mRNA JI876559.1 - 8 - 9 1240 61

View Full Product Details