Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhdc9
Entrez Gene ID
100766323
Gene Name
kelch domain containing 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614301: 168870 - 172359 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506858.2 XP_003506906.2 2 - 3 859 80
XM_007647391.1 XP_007645581.1 2 - 3 937 80
GenBank mRNA JI893897.1 - 1 - 2 80

View Full Product Details