Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lect2
Entrez Gene ID
100753512
Gene Name
leukocyte cell-derived chemotaxin 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613997: 292627 - 296793 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503736.1 XP_003503784.1 2 - 3 141 58

View Full Product Details