Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SFRP5
Entrez Gene ID
102173712
Gene Name
secreted frizzled related protein 5
Gene Aliases
-
Chromosome Location
Chr.26: 32696011 - 32700885 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JT671586.1 - 3 - 4 676 79

View Full Product Details