Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORN2
Entrez Gene ID
102179954
Gene Name
MORN repeat containing 2
Gene Aliases
-
Chromosome Location
Chr.11: 21051582 - 21060221 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686523.2 XP_005686580.1 4 - 5 446 86
XM_005686524.3 XP_005686581.1 3 - 4 377 86
GenBank mRNA JO453842.2 - 1 - 2 186 86
JO456326.2 - 1 - 2 186 86

View Full Product Details