Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REX1BD
Entrez Gene ID
102169273
Gene Name
required for excision 1-B domain containing
Gene Aliases
C7H19orf60
Chromosome Location
Chr.7: 104147257 - 104149926 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051444.1 XP_017906933.1 4 - 5 507 65
GenBank mRNA KA328658.1 - 2 - 3 350 65

View Full Product Details