Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HARBI1
Entrez Gene ID
102168752
Gene Name
harbinger transposase derived 1
Gene Aliases
-
Chromosome Location
Chr.15: 6407614 - 6416299 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690150.2 XP_005690207.1 2 - 3 919 61
GenBank mRNA JO591833.2 - 2 - 3 891 61

View Full Product Details