Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUS4L
Entrez Gene ID
102174088
Gene Name
dihydrouridine synthase 4 like
Gene Aliases
-
Chromosome Location
Chr.4: 71679994 - 71695870 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679131.3 XP_005679188.1 3 - 4 406 71
GenBank mRNA JO443816.2 - 4 - 5 576 71

View Full Product Details