Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM6
Entrez Gene ID
102176319
Gene Name
PR/SET domain 6
Gene Aliases
-
Chromosome Location
Chr.7: 79932917 - 80043784 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050681.1 XP_017906170.1 6 - 7 1864 94

View Full Product Details