Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCX
Entrez Gene ID
102184230
Gene Name
doublecortin
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189517: 32724033 - 32763971 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044095.1 XP_017899584.1 3 - 4 427 74

View Full Product Details