Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGR3
Entrez Gene ID
102180184
Gene Name
anterior gradient 3, protein disulphide isomerase family member
Gene Aliases
-
Chromosome Location
Chr.4: 95164713 - 95177409 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678999.3 XP_005679056.1 6 - 7 481 80

View Full Product Details