Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FGF11
Entrez Gene ID
102186718
Gene Name
fibroblast growth factor 11
Gene Aliases
-
Chromosome Location
Chr.19: 26926588 - 26933626 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064579.1 XP_017920068.1 2 - 3 2040 74
XM_018064580.1 XP_017920069.1 2 - 3 2040 74

View Full Product Details