Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL19
Entrez Gene ID
102176445
Gene Name
interleukin 19
Gene Aliases
-
Chromosome Location
Chr.16: 3986766 - 3996177 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690417.2 XP_005690474.1 3 - 4 341 66
XM_018060098.1 XP_017915587.1 3 - 4 341 66
XM_018060099.1 XP_017915588.1 3 - 4 342 66
XM_018060102.1 XP_017915591.1 3 - 4 323 66

View Full Product Details