The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005689730.3 | XP_005689787.1 | 2 - 3 | 945 | 90 |
GenBank mRNA | JO542256.2 | - | 2 - 3 | 773 | 90 |
JO584745.2 | - | 1 - 2 | 3277 | 90 |