Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNA15
Entrez Gene ID
102177052
Gene Name
G protein subunit alpha 15
Gene Aliases
-
Chromosome Location
Chr.7: 89884033 - 89904168 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050801.1 XP_017906290.1 5 - 6 1115 76

View Full Product Details