Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
QDPR
Entrez Gene ID
102176029
Gene Name
quinoid dihydropteridine reductase
Gene Aliases
-
Chromosome Location
Chr.6: 113029748 - 113049210 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049755.1 XP_017905244.1 3 - 4 339 78
GenBank mRNA JO420182.2 - 4 - 5 1398 78
KA338431.1 - 3 - 4 350 78

View Full Product Details