Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C14H8orf34
Entrez Gene ID
102174760
Gene Name
chromosome 14 C8orf34 homolog
Gene Aliases
-
Chromosome Location
Chr.14: 49386621 - 49724726 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689042.3 XP_005689099.2 7 - 8 1319 107

View Full Product Details