Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OXTR
Entrez Gene ID
102185124
Gene Name
oxytocin receptor
Gene Aliases
-
Chromosome Location
Chr.22: 17594775 - 17607957 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038465.1 XP_017893954.1 1 - 2 959 70

View Full Product Details