Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5DC3
Entrez Gene ID
102188832
Gene Name
5'-nucleotidase domain containing 3
Gene Aliases
-
Chromosome Location
Chr.5: 66140590 - 66203998 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048183.1 XP_017903672.1 10 - 11 1191 68
GenBank mRNA JO643822.2 - 2 - 3 103 68

View Full Product Details