Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RCOR2
Entrez Gene ID
102187746
Gene Name
REST corepressor 2
Gene Aliases
-
Chromosome Location
Chr.29: 42643114 - 42648322 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043196.1 XP_017898685.1 11 - 12 1942 55
GenBank mRNA JO624914.2 - 1 - 2 91 55

View Full Product Details